ENA379364443 Enamine

6-chloro-3-nitro-2-phenylpyridine

  • MDL number MFCD21790369
  • Cas Number : 1049706-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118879
Availability details for
Quantity
  • Availability
  • Add to Cart