ENA379364444 Enamine

3-phenylcyclopentan-1-amine hydrochloride

  • MDL number MFCD01735276
  • Cas Number : 56740-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118912
Availability details for
Quantity
  • Availability
  • Add to Cart