ENA379364446 Enamine

2,3-dihydro-1H-indole-6-carbonitrile

  • MDL number MFCD08544327
  • Cas Number : 15861-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118814

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118814
Availability details for
Quantity
  • Availability
  • Add to Cart