ENA379364468 Enamine

4-ethynyl-2,6-difluoroaniline

  • MDL number MFCD08703479
  • Cas Number : 753501-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118827
Availability details for
Quantity
  • Availability
  • Add to Cart