ENA379364477 Enamine

(6-chloro-2-methylpyridin-3-yl)methanol

  • MDL number MFCD13689158
  • Cas Number : 1093879-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119026
Availability details for
Quantity
  • Availability
  • Add to Cart