ENA379364487 Enamine

4-(1-hydroxyethyl)-2,6-dimethoxyphenol

  • MDL number MFCD00238585
  • Cas Number : 33900-62-8
  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(1-hydroxyethyl)-2,6-dimethoxyphenol
Inchi Code 1S/C10H14O4/c1-6(11)7-4-8(13-2)10(12)9(5-7)14-3/h4-6,11-12H,1-3H3
Inchi Key DDDDTTULEBRRJZ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-119110

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119110
Availability details for
Quantity
  • Availability
  • Add to Cart