ENA379364500 Enamine

2-amino-5-(morpholin-4-yl)benzamide

  • MDL number MFCD12406908
  • Cas Number : 93246-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26189

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26189
Availability details for
Quantity
  • Availability
  • Add to Cart