ENA379364501 Enamine

2-bromo-N-(3,4-dihydro-2H-1-benzopyran-4-yl)-3-methylbutanamide

  • MDL number MFCD11547037
  • Cas Number : 1097794-77-8
  • Molecular Weight : 312.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-29127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29127
Availability details for
Quantity
  • Availability
  • Add to Cart