ENA379364507 Enamine

2-cyano-4-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD11650639
  • Cas Number : 1099632-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40111

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40111
Availability details for
Quantity
  • Availability
  • Add to Cart