ENA379364532 Enamine

2-(1H-1,2,3-triazol-1-yl)aniline

  • MDL number MFCD09047488
  • Cas Number : 15213-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57989

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57989
Availability details for
Quantity
  • Availability
  • Add to Cart