ENA379364539 Enamine

4-(chloromethyl)oxane

  • MDL number MFCD16745652
  • Cas Number : 863324-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62685

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62685
Availability details for
Quantity
  • Availability
  • Add to Cart