ENA379364544 Enamine

2-phenylpropanedial

  • MDL number MFCD01366394
  • Cas Number : 26591-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65259

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65259
Availability details for
Quantity
  • Availability
  • Add to Cart