ENA379364545 Enamine

1-methyl-1H-1,3-benzodiazole-5-carbaldehyde

  • MDL number MFCD09261052
  • Cas Number : 279226-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65721

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-65721
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart