ENA379364549 Enamine

4H,5H,6H,7H,8H-thieno[3,2-c]azepin-4-one

  • MDL number MFCD19688417
  • Cas Number : 4751-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66574
Availability details for
Quantity
  • Availability
  • Add to Cart