ENA379364576 Enamine

2-chloro-N-[3-(dimethylamino)propyl]acetamide hydrochloride

  • MDL number MFCD18426455
  • Cas Number : 42951-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93179

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93179
Availability details for
Quantity
  • Availability
  • Add to Cart