ENA379364582 Enamine

ethyl 2-[2-(4-chlorophenyl)acetamido]acetate

  • MDL number MFCD01131964
  • Cas Number : 313067-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118862

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118862
Availability details for
Quantity
  • Availability
  • Add to Cart