ENA379364596 Enamine

4,5,6,7-tetrahydro-1H-indazole-7-carboxylic acid

  • MDL number MFCD22578759
  • Cas Number : 1423035-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98912
Availability details for
Quantity
  • Availability
  • Add to Cart