ENA387772231 Enamine

3-(1,3-thiazol-2-yloxy)pyridine

  • MDL number MFCD22628346
  • Cas Number : 1182041-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112439
Availability details for
Quantity
  • Availability
  • Add to Cart