ENA387772233 Enamine

methyl 4-nitro-3-(oxan-4-yl)butanoate

  • MDL number MFCD22628355
  • Cas Number : 1423024-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117560

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117560
Availability details for
Quantity
  • Availability
  • Add to Cart