ENA387772234 Enamine

tert-butyl N-(1,2,4-oxadiazol-3-yl)carbamate

  • MDL number MFCD22628356
  • Cas Number : 39512-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117659
Availability details for
Quantity
  • Availability
  • Add to Cart