ENA387772237 Enamine

2-(4-iodophenyl)pyrrolidine hydrochloride

  • MDL number MFCD22628361
  • Cas Number : 1423027-87-5
  • Molecular Weight : 309.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118107

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118107
Availability details for
Quantity
  • Availability
  • Add to Cart