ENA387772240 Enamine

2-(2,2-difluoro-3-oxo-3,4-dihydro-2H-1,4-benzoxazin-4-yl)acetic acid

  • MDL number MFCD22628366
  • Cas Number : 1423027-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118741

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118741
Availability details for
Quantity
  • Availability
  • Add to Cart