ENA387772241 Enamine

2-(3,5-dichlorophenyl)-2-(methylamino)acetic acid hydrochloride

  • MDL number MFCD22628367
  • Cas Number : 1423023-99-7
  • Molecular Weight : 270.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-118765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118765
Availability details for
Quantity
  • Availability
  • Add to Cart