ENA387772245 Enamine

4-tert-butyl-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD22628371
  • Cas Number : 1423029-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118991

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118991
Availability details for
Quantity
  • Availability
  • Add to Cart