ENA387772247 Enamine

2-(3-bromophenoxy)pyrazine hydrochloride

  • MDL number MFCD22628376
  • Cas Number : 1423027-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119099

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119099
Availability details for
Quantity
  • Availability
  • Add to Cart