ENA387772248 Enamine

3-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}propan-1-amine hydrochloride

  • MDL number MFCD22628377
  • Cas Number : 1423031-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119100
Availability details for
Quantity
  • Availability
  • Add to Cart