ENA387772251 Enamine

N-(5-methyl-1,3-thiazolidin-2-ylidene)methanamine

  • Cas Number : 21018-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119169
Availability details for
Quantity
  • Availability
  • Add to Cart