ENA387772256 Enamine

1-[(5-cyclohexyl-1,2,4-oxadiazol-3-yl)methyl]piperazine hydrochloride

  • MDL number MFCD22628391
  • Cas Number : 1423033-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119197

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119197
Availability details for
Quantity
  • Availability
  • Add to Cart