ENA387772258 Enamine

4-chloro-2-(1,3-dioxolan-2-yl)-1,3-thiazole

  • MDL number MFCD22628393
  • Cas Number : 1423025-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119231

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119231
Availability details for
Quantity
  • Availability
  • Add to Cart