ENA387772259 Enamine

3-(5-hydroxypyridin-3-yl)prop-2-enoic acid

  • MDL number MFCD22628396
  • Cas Number : 1807940-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 243...245
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119250

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119250
Availability details for
Quantity
  • Availability
  • Add to Cart