ENA387772260 Enamine

N-[(3-cycloheptyl-1,2,4-oxadiazol-5-yl)methyl]cyclopropanamine hydrochloride

  • MDL number MFCD22628397
  • Cas Number : 1423027-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119254

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119254
Availability details for
Quantity
  • Availability
  • Add to Cart