ENA387772262 Enamine

methyl 3-(2-chloroacetyl)benzoate

  • MDL number MFCD22628400
  • Cas Number : 177328-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-119259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119259
Availability details for
Quantity
  • Availability
  • Add to Cart