ENA387772263 Enamine

N-{[5-(propan-2-yl)-1,2,4-oxadiazol-3-yl]methyl}-N-(pyrrolidin-3-yl)acetamide hydrochloride

  • MDL number MFCD22628401
  • Cas Number : 1423034-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-119286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119286
Availability details for
Quantity
  • Availability
  • Add to Cart