ENA387772264 Enamine

3-(5-chloro-1-methyl-1H-imidazol-2-yl)-4,4,5,5,5-pentafluoro-3-hydroxypentanoic acid hydrochloride

  • MDL number MFCD22628402
  • Cas Number : 1423028-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119311
Availability details for
Quantity
  • Availability
  • Add to Cart