ENA387772266 Enamine

3-benzylimidazolidin-4-one hydrochloride

  • MDL number MFCD22628404
  • Cas Number : 114981-13-4
  • Molecular Weight : 212.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 164-169
Iupac Name 3-benzyl-4-imidazolidinone hydrochloride
Inchi Code 1S/C10H12N2O.ClH/c13-10-6-11-8-12(10)7-9-4-2-1-3-5-9;/h1-5,11H,6-8H2;1H
Inchi Key CFHCOTNUNBMOBE-UHFFFAOYSA-N

Description

Melting Point 164-169
Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-119321

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-119321
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart