ENA387772268 Enamine

(1R)-1-(4-methanesulfonylphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD22628406
  • Cas Number : 1415257-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119360
Availability details for
Quantity
  • Availability
  • Add to Cart