ENA387772274 Enamine

2-(4-amino-5-methyl-1H-pyrazol-1-yl)ethan-1-ol dihydrochloride

  • MDL number MFCD22628413
  • Cas Number : 1423033-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119425
Availability details for
Quantity
  • Availability
  • Add to Cart