ENA387772276 Enamine

tert-butyl N-[(5-acetyl-6-methylpyridin-2-yl)methyl]carbamate

  • MDL number MFCD22628416
  • Cas Number : 1423030-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72...74
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119450

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119450
Availability details for
Quantity
  • Availability
  • Add to Cart