ENA387772279 Enamine

2-[(dimethylamino)methyl]pyridin-4-amine dihydrochloride

  • MDL number MFCD22628421
  • Cas Number : 1423032-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119562

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119562
Availability details for
Quantity
  • Availability
  • Add to Cart