ENA387772280 Enamine

2-amino-N-[1-(pyridin-3-yl)ethyl]hexanamide hydrochloride

  • MDL number MFCD22628422
  • Cas Number : 1423023-82-8
  • Molecular Weight : 271.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119564
Availability details for
Quantity
  • Availability
  • Add to Cart