ENA387772289 Enamine

N-[(2-methylphenyl)methyl]pyrrolidine-2-carboxamide hydrochloride

  • MDL number MFCD22628437
  • Cas Number : 1423025-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119698

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119698
Availability details for
Quantity
  • Availability
  • Add to Cart