ENA387772293 Enamine

3-amino-2-methyl-2-[(thiophen-2-yl)methyl]propan-1-ol hydrochloride

  • MDL number MFCD22628443
  • Cas Number : 1423028-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119716

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119716
Availability details for
Quantity
  • Availability
  • Add to Cart