ENA387772299 Enamine

2-amino-2-[4-(trifluoromethyl)phenyl]ethan-1-ol hydrochloride

  • MDL number MFCD22628451
  • Cas Number : 1423032-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119823

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119823
Availability details for
Quantity
  • Availability
  • Add to Cart