ENA387772301 Enamine

3-amino-2-[(furan-2-yl)methyl]-2-methylpropanoic acid hydrochloride

  • MDL number MFCD22628454
  • Cas Number : 1423028-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119835

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119835
Availability details for
Quantity
  • Availability
  • Add to Cart