ENA387772304 Enamine

1-[5-(difluoromethyl)-1,2,4-oxadiazol-3-yl]ethan-1-amine hydrochloride

  • MDL number MFCD22628459
  • Cas Number : 1423023-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119882

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119882
Availability details for
Quantity
  • Availability
  • Add to Cart