ENA387772306 Enamine

methyl 4-cyclopropyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD22628461
  • Cas Number : 1423028-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119897
Availability details for
Quantity
  • Availability
  • Add to Cart