ENA387772307 Enamine

1-(3-tert-butyl-1,2,4-oxadiazol-5-yl)cyclohexan-1-amine hydrochloride

  • MDL number MFCD22628464
  • Cas Number : 1423025-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119907
Availability details for
Quantity
  • Availability
  • Add to Cart