ENA387772308 Enamine

2-(3-bromo-2-methoxyphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD22628465
  • Cas Number : 1423025-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119916
Availability details for
Quantity
  • Availability
  • Add to Cart