ENA387772315 Enamine

(1R)-1-(3-nitrophenyl)propan-1-amine hydrochloride

  • MDL number MFCD22628472
  • Cas Number : 1423015-67-1
  • Molecular Weight : 216.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R)-1-(3-nitrophenyl)-1-propanamine hydrochloride
Inchi Code 1S/C9H12N2O2.ClH/c1-2-9(10)7-4-3-5-8(6-7)11(12)13;/h3-6,9H,2,10H2,1H3;1H/t9-;/m1./s1
Inchi Key DOUHSTOUMBJFNC-SBSPUUFOSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-119940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119940
Availability details for
Quantity
  • Availability
  • Add to Cart