ENA387772324 Enamine

methyl[2-(propane-2-sulfonyl)ethyl]amine hydrochloride

  • MDL number MFCD22628482
  • Cas Number : 1423029-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119981
Availability details for
Quantity
  • Availability
  • Add to Cart